Merck
Characterization of natural aryl hydrocarbon receptor agonists from cassia seed and rosemary.
Molecules (Basel, Switzerland) (2014-04-22)
Yoshiaki Amakura, Morio Yoshimura, Masashi Takaoka, Haruka Toda, Tomoaki Tsutsumi, Rieko Matsuda, Reiko Teshima, Masafumi Nakamura, Hiroshi Handa, Takashi Yoshida
RESUMO
Many recent studies have suggested that activation of the aryl hydrocarbon receptor (AhR) reduces immune responses, thus suppressing allergies and autoimmune diseases. In our continuing study on natural AhR agonists in foods, we examined the influence of 37 health food materials on the AhR using a reporter gene assay, and found that aqueous ethanol extracts of cassia seed and rosemary had particularly high AhR activity. To characterize the AhR-activating substances in these samples, the chemical constituents of the respective extracts were identified. From an active ethyl acetate fraction of the cassia seed extract, eight aromatic compounds were isolated. Among these compounds, aurantio-obtusin, an anthraquinone, elicited marked AhR activation. Chromatographic separation of an active ethyl acetate fraction of the rosemary extract gave nine compounds. Among these compounds, cirsimaritin induced AhR activity at 10-10² μM, and nepitrin and homoplantagenin, which are flavone glucosides, showed marked AhR activation at 10-10³ μM.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Homoplantaginin, ≥98% (HPLC)