Merck
  • Concentration and congener pattern of polychlorinated biphenyls in blubber and liver of Hubbs' beaked whale (Mesoplodon carlhubbsi), using a sulfoxide and an Ag-ION solid phase extraction cartridge as a simplified cleanup technique for biological samples.
Concentration and congener pattern of polychlorinated biphenyls in blubber and liver of Hubbs' beaked whale (Mesoplodon carlhubbsi), using a sulfoxide and an Ag-ION solid phase extraction cartridge as a simplified cleanup technique for biological samples.
Marine pollution bulletin (2016-10-07)
Katsunori Anezaki, Ayaka Matsuda, Takashi Matsuishi
RESUMO
We performed the first known study of polychlorinated biphenyls (PCBs) concentrations and patterns in the blubber and liver of a Hubbs' beaked whale. Samples were pretreated with Supelclean™ sulfoxide and Discovery® Ag-ION solid phase extraction cartridges to remove whale oil. PCB concentrations in the blubber and liver were 13,000 and 7300ng/g lipid, respectively. Highly poisonous congeners such as dioxin-like (DL) PCBs tended to accumulate in the liver. The toxic equivalents (TEQ) of DL-PCBs in the liver (740pg-TEQ/g lipid) were higher than those in the blubber (74pg-TEQ/g lipid). The blubber and liver samples showed that hexachlorinated biphenyls were dominant among homologues, and PCB-153 was dominant among congeners. Several congeners accumulated disproportionately in the blubber and the liver (PCB-28, 52, 74, 99, and 118), while others did not persist (PCB-31, 70, and 110). This indicates that PCBs are metabolized differently according to their specific composition.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Discovery® Ag-Ion SPE Tube, bed wt. 750 mg, volume 6 mL, pk of 30