Merck
  • Analysis of Chromatin Opening in Heterochromatic Non-Small Cell Lung Cancer Tumor-Initiating Cells in Relation to DNA-Damaging Antitumor Treatment.
Analysis of Chromatin Opening in Heterochromatic Non-Small Cell Lung Cancer Tumor-Initiating Cells in Relation to DNA-Damaging Antitumor Treatment.
International journal of radiation oncology, biology, physics (2017-11-07)
Mina Eriksson, Petra Hååg, Beata Brzozowska, Magdalena Lipka, Halina Lisowska, Rolf Lewensohn, Andrzej Wojcik, Kristina Viktorsson, Lovisa Lundholm
RESUMO
We previously reported that sphere-forming non-small cell lung cancer (NSCLC) tumor-initiating cells (TICs) have an altered activation of DNA damage response- and repair proteins and are refractory to DNA-damaging treatments. We analyzed whether chromatin organization plays a role in the observed refractoriness. Bulk cells and TICs from the NSCLC H23 and H1299 cell lines were examined using cell viability, clonogenic survival, Western blot, short interfering RNA analysis, and micronucleus assay. NSCLC TICs displayed elevated heterochromatin markers trimethylated lysine 9 of histone H3 and heterochromatin protein 1γ relative to bulk cells and reduced cell viability upon histone deacetylase inhibition (HDACi). Vorinostat and trichostatin A increased the euchromatin markers acetylated lysine 9/14 of histone H3 and lysine 8 of histone H4, and HDACi pretreatment increased the phosphorylation of the DNA damage response proteins ataxia telangiectasia mutated and DNA-dependent protein kinase, catalytic subunit, upon irradiation in TICs. HDACi sensitized TICs to cisplatin and to some extent to ionizing irradiation. The protectiveness of a dense chromatin structure was indicated by an enhanced frequency of micronuclei in TICs following irradiation, after knockdown of heterochromatin protein 1γ. Although confirmatory studies in additional NSCLC model systems and with respect to analyses of other DNA damage response proteins are needed, our data point toward a heterochromatic structure of NSCLC TICs, such that HDACi can sensitize TICs to DNA damage.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Sirtinol, ≥95% (HPLC)