Merck
  • The Drosophila Gr28bD product is a non-specific cation channel that can be used as a novel thermogenetic tool.
The Drosophila Gr28bD product is a non-specific cation channel that can be used as a novel thermogenetic tool.
Scientific reports (2018-01-19)
Aditi Mishra, Autoosa Salari, Benton R Berigan, Kayla C Miguel, Marzie Amirshenava, Abbey Robinson, Benjamin C Zars, Jenna L Lin, Lorin S Milescu, Mirela Milescu, Troy Zars
RESUMO
Extrinsic control of single neurons and neuronal populations is a powerful approach for understanding how neural circuits function. Adding new thermogenetic tools to existing optogenetic and other forms of intervention will increase the complexity of questions that can be addressed. A good candidate for developing new thermogenetic tools is the Drosophila gustatory receptor family, which has been implicated in high-temperature avoidance behavior. We examined the five members of the Gr28b gene cluster for temperature-dependent properties via three approaches: biophysical characterization in Xenopus oocytes, functional calcium imaging in Drosophila motor neurons, and behavioral assays in adult Drosophila. Our results show that Gr28bD expression in Xenopus oocytes produces a non-specific cationic current that is activated by elevated temperatures. This current is non-inactivating and non-voltage dependent. When expressed in Drosophila motor neurons, Gr28bD can be used to change the firing pattern of individual cells in a temperature-dependent fashion. Finally, we show that pan-neuronal or motor neuron expression of Gr28bD can be used to alter fruit fly behavior with elevated temperatures. Together, these results validate the potential of the Gr28bD gene as a founding member of a new class of thermogenetic tools.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
LCS-1, ≥98% (HPLC)