Merck
  • Evidence for C1q-mediated crosslinking of CD33/LAIR-1 inhibitory immunoreceptors and biological control of CD33/LAIR-1 expression.
Evidence for C1q-mediated crosslinking of CD33/LAIR-1 inhibitory immunoreceptors and biological control of CD33/LAIR-1 expression.
Scientific reports (2017-03-23)
Myoungsun Son, Betty Diamond, Bruce T Volpe, Cynthia B Aranow, Meggan C Mackay, Frances Santiago-Schwarz
RESUMO
C1q collagen-like region (CLR) engaging and activating the LAIR-1 inhibitory immunoreceptor represents a non-complement mechanism for maintaining immune quiescence. Given the binding promiscuity of C1q's globular region (gC1q), we hypothesized that C1q concurrently associates with distinct inhibitory immunoreceptors to produce C1q-mediated modulatory networking. Like LAIR-1, CD33 inhibitory immunoreceptors are highly expressed on monocytes. Binding CD33 restricts cell activation/differentiation; however, natural ligands for CD33 remain elusive. CD33 has IgC2-like domains potentially recognized by gC1q. Thus, we asked whether C1q binds to CD33 and if C1q mediates CD33/LAIR-1 crosslinking. Our findings demonstrate that C1q and gC1q interact with CD33 to activate its inhibitory motifs, while CLR does not. Whole C1q is required to crosslink CD33 and LAIR-1 and concurrently activate CD33/LAIR-1 inhibitory motifs. While C1q binds CD33C2 domains, decreased C1q-CD33 interactions resulting from sialic acid masking of CD33C2 domains suggests a process for regulating C1q-CD33 activity. Consistent with defective self-tolerance, CD33/LAIR-1 expression is reduced in systemic lupus erythematosus (SLE) myelomonocytes. The anti-inflammatory cytokine M-CSF, but not DC growth factors, sustains CD33/LAIR-1 expression on both healthy and SLE cells suggesting further biological control of C1q-CD33/LAIR-1 processes.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anti-LAIR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution