Merck
  • Structure guided design and kinetic analysis of highly potent benzimidazole inhibitors targeting the PDEδ prenyl binding site.
Structure guided design and kinetic analysis of highly potent benzimidazole inhibitors targeting the PDEδ prenyl binding site.
Journal of medicinal chemistry (2014-06-03)
Gunther Zimmermann, Carsten Schultz-Fademrecht, Philipp Küchler, Sandip Murarka, Shehab Ismail, Gemma Triola, Peter Nussbaumer, Alfred Wittinghofer, Herbert Waldmann
RESUMO
K-Ras is one of the most frequently mutated signal transducing human oncogenes. Ras signaling activity requires correct cellular localization of the GTPase. The spatial organization of K-Ras is controlled by the prenyl binding protein PDEδ, which enhances Ras diffusion in the cytosol. Inhibition of the Ras-PDEδ interaction by small molecules impairs Ras localization and signaling. Here we describe in detail the identification and structure guided development of Ras-PDEδ inhibitors targeting the farnesyl binding pocket of PDEδ with nanomolar affinity. We report kinetic data that characterize the binding of the most potent small molecule ligands to PDEδ and prove their binding to endogenous PDEδ in cell lysates. The PDEδ inhibitors provide promising starting points for the establishment of new drug discovery programs aimed at cancers harboring oncogenic K-Ras.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Deltarasin trihydrochloride, ≥98% (HPLC)