- Structural basis for maintenance of bacterial outer membrane lipid asymmetry.
Structural basis for maintenance of bacterial outer membrane lipid asymmetry.
Nature microbiology (2017-10-19)
Javier Abellón-Ruiz, Shreyas S Kaptan, Arnaud Baslé, Beatrice Claudi, Dirk Bumann, Ulrich Kleinekathöfer, Bert van den Berg
PMID29038444
RESUMO
The Gram-negative bacterial outer membrane (OM) is a unique bilayer that forms an efficient permeation barrier to protect the cell from noxious compounds
MATERIAIS
Número do produto
Marca
Descrição do produto