Merck
  • LC-MS/MS method for the quantification of masitinib in RLMs matrix and rat urine: application to metabolic stability and excretion rate.
LC-MS/MS method for the quantification of masitinib in RLMs matrix and rat urine: application to metabolic stability and excretion rate.
Chemistry Central journal (2017-12-24)
Sawsan M Amer, Adnan A Kadi, Hany W Darwish, Mohamed W Attwa
RESUMO
Masitinib (MST) is a selective tyrosine kinase inhibitor. Validated liquid chromatography tandem mass spectrometric method (LC-MS/MS) was developed for the quantification of MST in rat liver microsomes (RLMs) matrix. The developed method was applied to metabolic stability and excretion rate studies. Reversed phase liquid chromatography was used for resolution of MST and bosutinib (IS) using C
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Bosutinib, ≥98% (HPLC)