Merck
Identification of a testicular odorant receptor mediating human sperm chemotaxis.
Science (New York, N.Y.) (2003-03-29)
Marc Spehr, Gunter Gisselmann, Alexandra Poplawski, Jeffrey A Riffell, Christian H Wetzel, Richard K Zimmer, Hanns Hatt
RESUMO
Although it has been known for some time that olfactory receptors (ORs) reside in spermatozoa, the function of these ORs is unknown. Here, we identified, cloned, and functionally expressed a previously undescribed human testicular OR, hOR17-4. With the use of ratiofluorometric imaging, Ca2+ signals were induced by a small subset of applied chemical stimuli, establishing the molecular receptive fields for the recombinantly expressed receptor in human embryonic kidney (HEK) 293 cells and the native receptor in human spermatozoa. Bourgeonal was a powerful agonist for both recombinant and native receptor types, as well as a strong chemoattractant in subsequent behavioral bioassays. In contrast, undecanal was a potent OR antagonist to bourgeonal and related compounds. Taken together, these results indicate that hOR17-4 functions in human sperm chemotaxis and may be a critical component of the fertilization process.