Merck
Structure-based design of aliskiren, a novel orally effective renin inhibitor.
Biochemical and biophysical research communications (2003-08-21)
Jeanette M Wood, Jürgen Maibaum, Joseph Rahuel, Markus G Grütter, Nissim-Claude Cohen, Vittorio Rasetti, Heinrich Rüger, Richard Göschke, Stefan Stutz, Walter Fuhrer, Walter Schilling, Pascal Rigollier, Yasuchika Yamaguchi, Frederic Cumin, Hans-Peter Baum, Christian R Schnell, Peter Herold, Robert Mah, Chris Jensen, Eoin O'Brien, Alice Stanton, Martin P Bedigian
RESUMO
Hypertension is a major risk factor for cardiovascular diseases such as stroke, myocardial infarction, and heart failure, the leading causes of death in the Western world. Inhibitors of the renin-angiotensin system (RAS) have proven to be successful treatments for hypertension. As renin specifically catalyses the rate-limiting step of the RAS, it represents the optimal target for RAS inhibition. Several peptide-like renin inhibitors have been synthesized previously, but poor pharmacokinetic properties meant that these compounds were not clinically useful. We employed a combination of molecular modelling and crystallographic structure analysis to design renin inhibitors lacking the extended peptide-like backbone of earlier inhibitors, for improved pharmacokinetic properties. This led to the discovery of aliskiren, a highly potent and selective inhibitor of human renin in vitro, and in vivo; once-daily oral doses of aliskiren inhibit renin and lower blood pressure in sodium-depleted marmosets and hypertensive human patients. Aliskiren represents the first in a novel class of renin inhibitors with the potential for treatment of hypertension and related cardiovascular diseases.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Aliskiren hemifumarate, ≥95% (HPLC)