Merck
Specific Pathogen Recognition by Multiple Innate Immune Sensors in an Invertebrate.
Frontiers in immunology (2017-10-21)
Guillaume Tetreau, Silvain Pinaud, Anaïs Portet, Richard Galinier, Benjamin Gourbal, David Duval
RESUMO
Detection of pathogens by all living organisms is the primary step needed to implement a coherent and efficient immune response. This implies a mediation by different soluble and/or membrane-anchored proteins related to innate immune receptors called PRRs (pattern-recognition receptors) to trigger immune signaling pathways. In most invertebrates, their roles have been inferred by analogy to those already characterized in vertebrate homologs. Despite the induction of their gene expression upon challenge and the presence of structural domains associated with the detection of pathogen-associated molecular patterns in their sequence, their exact role in the induction of immune response and their binding capacity still remain to be demonstrated. To this purpose, we developed a fast interactome approach, usable on any host-pathogen couple, to identify soluble proteins capable of directly or indirectly detecting the presence of pathogens. To investigate the molecular basis of immune recognition specificity, different pathogens (Gram-positive bacterium,
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Piperazine diacrylamide, For acrylamide gel electrophoresis