Merck
Metabolic Comparison of Dorsal
Analytical methods : advancing methods and applications (2017-10-25)
Rosemary M Onjiko, David O Plotnick, Sally A Moody, Peter Nemes
RESUMO
Single-cell mass spectrometry (MS) empowers the characterization of metabolomic changes as cells differentiate to different tissues during early embryogenesis. Using whole-cell dissection and capillary electrophoresis electrospray ionization (CE-ESI) MS, we recently uncovered metabolic cell-to-cell differences in the 8- and 16-cell embryo of the South African clawed frog (
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Calcium nitrite solution, 30 wt. % in H2O