- Anatomical distribution of primary amine oxidase activity in four adipose depots and plasma of severely obese women with or without a dysmetabolic profile.
Anatomical distribution of primary amine oxidase activity in four adipose depots and plasma of severely obese women with or without a dysmetabolic profile.
Journal of physiology and biochemistry (2016-10-22)
Christian Carpéné, Francisco Les, Mounia Hasnaoui, Simon Biron, Picard Marceau, Denis Richard, Jean Galitzky, Denis R Joanisse, Pascale Mauriège
PMID27766585
RESUMO
Semicarbazide-sensitive amine oxidase (SSAO), identical to primary amine oxidase or vascular adhesion protein-1, is a membrane enzyme that generates hydrogen peroxide. SSAO is highly expressed at the adipocyte surface, and its plasma levels increase with type 2 diabetes. Since visceral adipose tissue (AT) is more tightly associated with obesity complications than subcutaneous (SC) abdominal fat, we compared SSAO activity in plasma and 4 distinct AT locations in 48 severely obese women (body mass index (BMI), averaging 54 ± 11 kg/m
MATERIAIS