Merck
  • DJ-1 regulating PI3K-Nrf2 signaling plays a significant role in bibenzyl compound 20C-mediated neuroprotection against rotenone-induced oxidative insult.
DJ-1 regulating PI3K-Nrf2 signaling plays a significant role in bibenzyl compound 20C-mediated neuroprotection against rotenone-induced oxidative insult.
Toxicology letters (2017-03-02)
Xiao-Ling Zhang, Yu-He Yuan, Qian-Hang Shao, Zhen-Zhen Wang, Cheng-Gen Zhu, Jian-Gong Shi, Kai-Li Ma, Xu Yan, Nai-Hong Chen
RESUMO
Oxidative stress is thought to be involved in the development of Parkinson's disease (PD). We previously reported that 20C, a bibenzyl compound isolated from Gastrodia elata, possesses antioxidative properties, but its in-depth molecular mechanisms against rotenone-induced neurotoxicity remains unknown. Recent studies indicate that without intact DJ-1, nuclear factor erythroid 2-related factor (Nrf2) protein becomes unstable, and the activity of Nrf2-mediated downstream antioxidant enzymes are thereby suppressed. In this study, we showed that 20C clearly protected PC12 and SH-SY5Y cells against rotenone-induced oxidative injury. Furthermore, 20C markedly up-regulated the levels of DJ-1, which in turn activated phosphoinositide-3-kinase (PI3K)/Akt signaling and inhibited glycogen synthase kinase 3β (GSK3β) activation, eventually promoted the nuclear translocation of Nrf2 and induced the expression of hemeoxygenase-1 (HO-1). The antioxidant effects of 20C could be partially blocked by ShRNA-mediated knockdown of DJ-1 and inhibition of the PI3K/Akt pathways with Akt1/2 kinase inhibitor, respectively. Conclusively, our findings confirm that DJ-1 is necessary for 20C-mediated protection against rotenone-induced oxidative damage, at least in part, by activating PI3K/Akt signaling, and subsequently enhancing the nuclear accumulation of Nrf2. The findings from our investigation suggest that 20C should be developed as a novel candidate for alleviating the consequences of PD in the future.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
MISSION® esiRNA, targeting human PARK7
Preço e disponibilidade não estão disponíveis no momento.