Merck
Synthesis and complexation of superbulky imidazolium-2-dithiocarboxylate ligands.
Dalton transactions (Cambridge, England : 2003) (2017-07-01)
Tomás F Beltrán, Guillermo Zaragoza, Lionel Delaude
RESUMO
The superbulky N-heterocyclic carbenes (NHCs) 2,6-bis(diphenylmethyl)-4-methylimidazol-2-ylidene (IDip*Me) and its 4-methoxy analogue (IDip*OMe) reacted instantaneously with carbon disulfide to afford the corresponding imidazolium-2-dithiocarboxylate zwitterions in high yields. These new dithiolate ligands were fully characterized and their coordination chemistry toward common Re(i) and Ru(ii) metal sources was thoroughly investigated. Neutral [ReBr(CO)
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Bromopentacarbonylrhenium(I), 98%