Merck
  • Development and Validation of Ion Chromatography-Tandem Mass Spectrometry-Based Method for the Multiresidue Determination of Polar Ionic Pesticides in Food.
Development and Validation of Ion Chromatography-Tandem Mass Spectrometry-Based Method for the Multiresidue Determination of Polar Ionic Pesticides in Food.
Journal of agricultural and food chemistry (2017-04-08)
Stuart Adams, Jonathan Guest, Michael Dickinson, Richard J Fussell, Jonathan Beck, Frans Schoutsen
RESUMO
An extraction method using acidified methanol based on the quick polar pesticide (QuPPe) method using suppressed ion chromatography coupled to mass spectrometry was developed and validated for the direct analysis of polar pesticides, without the need for derivatization or ion pairing, in cereals and grapes. The method was robust, and results for glyphosate, aminomethyl phosphonic acid (AMPA), N-acetyl-AMPA, glufosinate, 3-methylphosphinicopropionic acid (3-MPPA), N-acetyl glufosinate, ethephon, chlorate, perchlorate, fosetyl aluminum, and phosphonic acid at three concentration levels (typically 0.01, 0.05, and 0.1 mg/kg) were compliant with SANTE/11945/2015 guideline method performance criteria. Cereal-based infant food proved to be a more challenging matrix and validated only for glyphosate, chlorate, and perchlorate at 0.005, 0.01, and 0.05 mg/kg. The developed method enables the multiresidue analysis of 12 ionic pesticides and relevant metabolites in a single analysis. Until now, the analysis of these compounds required several different single-residue methods using different chromatographic conditions. This multiresidue approach offers the possibility of more cost-effective and more efficient monitoring of polar ionic pesticides and contaminants that are of concern to food regulation bodies and consumers.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Clopyralid, PESTANAL®, analytical standard