Merck
  • Offline Solid-phase Extraction Large-volume Injection-Gas chromatography for the Analysis of Mineral Oil-saturated Hydrocarbons in Commercial Vegetable Oils.
Offline Solid-phase Extraction Large-volume Injection-Gas chromatography for the Analysis of Mineral Oil-saturated Hydrocarbons in Commercial Vegetable Oils.
Journal of oleo science (2017-08-11)
Lingling Liu, Hua Huang, Yanwen Wu, Bingning Li, Jie Ouyang
RESUMO
An offline solid-phase extraction (SPE) approach combined with a large-volume injection (LVI)-gas chromatography-flame ionization detector (LVI-GC-FID) is improved for routine analysis of mineral oil saturated hydrocarbons (MOSH) in vegetable oils. The key procedure of the method consists in using offline SPE columns for MOSH purification. The SPE column packed with 1% Ag-activated silica gel was used to separate MOSH from triglycerides and olefins in variety of vegetable oils. The eluent of MOSH fraction was only 3 mL and the concentration step was quick with little evaporation loss. The limit of quantification (LOQ) of the method was 2.5 mg/kg and the linearity ranged from 2 to 300 mg/kg. The accuracy was assessed by measuring the recoveries from spiked oil samples and was higher than 90%. Twenty-seven commercial vegetable oils were analyzed, and different levels of MOSH contamination were detected with the highest being 259.4 mg/kg. The results suggested that it is necessary to routinely detect mineral oil contamination in vegetable oils for food safety.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
1,3,5-Tri-tert-butylbenzene, 97%