Merck
Expression of Interleukin-26 is upregulated in inflammatory bowel disease.
World journal of gastroenterology (2017-08-31)
Makoto Fujii, Atsushi Nishida, Hirotsugu Imaeda, Masashi Ohno, Kyohei Nishino, Shigeki Sakai, Osamu Inatomi, Shigeki Bamba, Masahiro Kawahara, Tomoharu Shimizu, Akira Andoh
RESUMO
To investigate interleukin (IL)-26 expression in the inflamed mucosa of patients with inflammatory bowel disease (IBD) and the function of IL-26. Human colonic subepithelial myofibroblasts (SEMFs) were isolated from colon tissue surgically resected. The expression of IL-26 protein and its receptor complex was analyzed by immunohistochemistry. The gene expression induced by IL-26 was evaluated by real-time polymerase chain reaction. Intracellular signaling pathways were evaluated by immunoblotting and specific small interfering (si) RNA transfection. The mRNA and protein expression of IL-26 were significantly enhanced in the inflamed mucosa of patients with IBD. IL-26 receptor complex was expressed in colonic SEMFs These results suggest that IL-26 plays a role in the pathophysiology of IBD through induction of inflammatory mediators.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
MISSION® esiRNA, targeting human STAT1