Merck
Sirtuin 6 contributes to migration and invasion of osteosarcoma cells via the ERK1/2/MMP9 pathway.
FEBS open bio (2017-09-15)
Hang Lin, Yi Hao, Zhengxu Zhao, Yongjun Tong
RESUMO
Dysregulation of sirtuin 6 (SIRT6) is actively involved in tumor progression. High levels of SIRT6 have been associated with hepatocellular carcinoma and non-small cell lung cancer, and SIRT6 facilitates growth and metastasis of cancer cells. However, the clinical significance and biological function of SIRT6 are not known for osteosarcoma (OS). Here, we report that SIRT6 was notably overexpressed in OS tissues compared with non-cancerous specimens. The high level of SIRT6 was prominently correlated with malignant clinical parameters and poor prognosis of OS patients. SIRT6 was also up-regulated in OS cells. SIRT6 knockdown inhibited the invasion and migration of Saos-2 and U2OS cells
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
MISSION® esiRNA, targeting human MMP9