- Suppressors of Superoxide-H
Suppressors of Superoxide-H
Cell metabolism (2016-09-27)
Martin D Brand, Renata L S Goncalves, Adam L Orr, Leonardo Vargas, Akos A Gerencser, Martin Borch Jensen, Yves T Wang, Simon Melov, Carolina N Turk, Jason T Matzen, Victoria J Dardov, H Michael Petrassi, Shelly L Meeusen, Irina V Perevoshchikova, Heinrich Jasper, Paul S Brookes, Edward K Ainscow
PMID27667666
RESUMO
Using high-throughput screening we identified small molecules that suppress superoxide and/or H
MATERIAIS