Merck
  • Silver sulfadiazine-a new topical therapy for pseudomonas in burns: therapy of pseudomonas infection in burns
Silver sulfadiazine-a new topical therapy for pseudomonas in burns: therapy of pseudomonas infection in burns
Archives of Surgery (Chicago, Ill. : 1960) (1968)
Fox CL