Merck
  • Magnetic nanoparticles and high-speed countercurrent chromatography coupled in-line and using the same solvent system for separation of quercetin-3-O-rutinoside, luteoloside and astragalin from a Mikania micrantha extract.
Magnetic nanoparticles and high-speed countercurrent chromatography coupled in-line and using the same solvent system for separation of quercetin-3-O-rutinoside, luteoloside and astragalin from a Mikania micrantha extract.
Journal of chromatography. A (2017-06-13)
Juanqiang Wang, Shan Geng, Binghai Wang, Qian Shao, Yingtong Fang, Yun Wei
RESUMO
A new in-line method of magnetic nanoparticles (MNPs) coupled with high-speed countercurrent chromatography (HSCCC) using a same solvent system during the whole separation process was established to achieve the rapid separation of flavonoids from Mikania micrantha. The adsorption and desorption capacities of five different MNPs for flavonoid standards and Mikania micrantha crude extract were compared and the most suitable magnetic nanoparticle Fe
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
1,3-Diiodo-5,5-dimethylhydantoin, ≥96%