Merck
The A
Cancer letters (2016-10-30)
Christina Mølck, James Ryall, Laura M Failla, Janine L Coates, Jean-Marc Pascussi, Joan K Heath, Gregory Stewart, Frédéric Hollande
RESUMO
Adenosine is a multifaceted regulator of tumor progression. It modulates immune cell activity as well as acting directly on tumor cells. The A The A PSB-603 alters cellular metabolism in colorectal cancer cells and increases their sensitivity to chemotherapy. Although requiring more mechanistic insight into its A
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
PSB-603, ≥98% (HPLC)