Merck
  • A role for beta-cell-expressed G protein-coupled receptor 119 in glycemic control by enhancing glucose-dependent insulin release.
A role for beta-cell-expressed G protein-coupled receptor 119 in glycemic control by enhancing glucose-dependent insulin release.
Endocrinology (2007-02-10)
Zhi-Liang Chu, Robert M Jones, Hongmei He, Chris Carroll, Veronica Gutierrez, Annette Lucman, Molly Moloney, Hui Gao, Helen Mondala, Didier Bagnol, David Unett, Yin Liang, Keith Demarest, Graeme Semple, Dominic P Behan, James Leonard
RESUMO
Pancreatic beta-cell dysfunction is a hallmark event in the pathogenesis of type 2 diabetes. Injectable peptide agonists of the glucagon-like peptide 1 (GLP-1) receptor have shown significant promise as antidiabetic agents by virtue of their ability to amplify glucose-dependent insulin release and preserve pancreatic beta-cell mass. These effects are mediated via stimulation of cAMP through beta-cell GLP-1 receptors. We report that the Galpha(s)-coupled receptor GPR119 is largely restricted to insulin-producing beta-cells of pancreatic islets. Additionally, we show here that GPR119 functions as a glucose-dependent insulinotropic receptor. Unlike receptors for GLP-1 and other peptides that mediate enhanced glucose-dependent insulin release, GPR119 was suitable for the development of potent, orally active, small-molecule agonists. The GPR119-specific agonist AR231453 significantly increased cAMP accumulation and insulin release in both HIT-T15 cells and rodent islets. In both cases, loss of GPR119 rendered AR231453 inactive. AR231453 also enhanced glucose-dependent insulin release in vivo and improved oral glucose tolerance in wild-type mice but not in GPR119-deficient mice. Diabetic KK/A(y) mice were also highly responsive to AR231453. Orally active GPR119 agonists may offer significant promise as novel antihyperglycemic agents acting in a glucose-dependent fashion.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
AR231453, ≥98% (HPLC)