- Integration of Triboluminescent EuD
Integration of Triboluminescent EuD
ACS applied materials & interfaces (2017-01-27)
Anıl İncel, Mehtap Emirdag-Eanes, Colin D McMillen, Mustafa M Demir
PMID28125205
RESUMO
Lanthanide-based organometallic materials are well-known candidate triboluminescent (TL) materials that can show bright emission when a mechanical force is applied. These materials are usually in the form of crystalline powders, and it is often useful to integrate these samples into a polymer matrix in order to achieve processability, enabling coating from a solution/molten state or fabrication as a complex-shaped matrix. In this work, micrometer-sized europium tetrakis (dibenzoylmethide) triethylammonium (EuD
MATERIAIS
Número do produto
Marca
Descrição do produto