Merck
  • Inhaled corticosteroid related adrenal suppression detected by poor growth and reversed with ciclesonide.
Inhaled corticosteroid related adrenal suppression detected by poor growth and reversed with ciclesonide.
The Journal of asthma : official journal of the Association for the Care of Asthma (2016-06-11)
Brian S Liddell, John M Oberlin, Daniel P Hsu
RESUMO
This case series intends to highlight the association between decreased linear growth velocity and adrenal suppression in patients receiving inhaled corticosteroids for asthma. A potential treatment option is also discussed. Adrenal suppression secondary to inhaled corticosteroids has previously been reported and is often underrecognized. A decrease in linear height velocity has also been associated with inhaled corticosteroids. However, a decrease in height velocity has not been shown to predict adrenal suppression. This case series of 15 patients receiving inhaled corticosteroids for control of asthma were noted to have a decrease in linear growth velocity ultimately associated with adrenal suppression. A change in inhaled corticosteroid to ciclesonide led to recovery of adrenal function without impacting asthma control. Chart review from a pediatric pulmonology clinic was performed. Growth parameters and laboratory studies were recorded and analyzed. A mean decrease in height standard deviation score in the year prior to diagnosis of adrenal suppression was -0.59, -0.11, and -0.18, in pre-puberty, peri-puberty, and post-puberty patients, respectively. After ciclesonide therapy was initiated, a mean change in height standard deviation score of +0.40, +0.13, and -0.13, was noted for pre-puberty, peri-puberty, and post-puberty patients, respectively. A change in growth velocity of +5.3 cm/yr, +2.1 cm/yr, and -1.9 cm/yr, was noted for pre-puberty, peri-puberty, and post-puberty patients, respectively, after starting ciclesonide. Height velocity should be monitored closely during routine asthma visits to identify potential adrenal suppression associated with inhaled corticosteroids use. Ciclesonide is a good option for asthma treatment that allows for adrenal recovery.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Ciclesonide, ≥98% (HPLC)