Merck
CdS/CdSe Co-sensitized Solar Cells Based on Hierarchically Structured SnO2/TiO2 Hybrid Films.
Nanoscale research letters (2016-06-15)
Zeng Chen, Chaochao Wei, Shengjun Li, Chunli Diao, Wei Li, Wenping Kong, Zhenlong Zhang, Weifeng Zhang
RESUMO
SnO2 nanosheet-structured films were prepared on a fluorine-doped tin oxide (FTO) substrate using ZnO nanosheet as template. The as-prepared SnO2 nanosheets contained plenty of nano-voids and were generally vertical to the substrate. TiO2 nanoparticles were homogeneously deposited into the intervals between the SnO2 nanosheets to prepare a hierarchically structured SnO2/TiO2 hybrid film. The hybrid films were co-sensitized with CdS and CdSe quantum dots. The sensitized solar cells assembled with the SnO2/TiO2 hybrid film showed much higher photoelectricity conversion efficiency than the cells assembled with pure TiO2 films. The lifetime of photoinduced electron was also investigated through electrochemical impedance spectroscopy, which showed that the SnO2/TiO2 hybrid film electrode is as long as the TiO2 film electrode.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Nitrilotriacetic acid trisodium salt, Sigma Grade, ≥98%
Sigma-Aldrich
Ammonium hexafluorostannate, ≥99.99% trace metals basis