Merck
  • Erufosine increases RhoB expression in oral squamous carcinoma cells independent of its tumor suppressive mode of action - a short report.
Erufosine increases RhoB expression in oral squamous carcinoma cells independent of its tumor suppressive mode of action - a short report.
Cellular oncology (Dordrecht) (2016-11-05)
Shariq S Ansari, Nurullah Akgün, Martin R Berger
RESUMO
Recently, we found that erufosine (erucylphospho-N,N,N trimethylpropylammonium) can induce up-regulation of RhoB expression in oral squamous carcinoma (OSCC) cells, thereby hinting at a tumor suppressive role. Therefore, we aimed to evaluate the role of RhoB in the tumor suppressive mode of action of erufosine on OSCC cells. Anti-proliferative effects of erufosine were determined in HN-5 and FaDu OSCC-derived cells using a MTT assay. RhoB up-regulation was detected using microarray and qRT-PCR-based expression assays at IC We found that after erufosine treatment of HN-5 and FaDu cells for 24, 48 and 72 h the IC Our data show that erufosine can cause up-regulation of RhoB expression in OSCC cells. Combining erufosine treatment with siRNA-mediated RhoB knockdown did, however, not reveal a role of RhoB in its tumor suppressive mode of action.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
MISSION® esiRNA, targeting human RHOB