Merck
Magnetically driven nanoparticles:
Contrast media & molecular imaging (2017-01-05)
Mariarosaria De Simone, Daniele Panetta, Emilia Bramanti, Cristiana Giordano, Maria C Salvatici, Lisa Gherardini, Arianna Menciassi, Silvia Burchielli, Caterina Cinti, Piero A Salvadori
RESUMO
Superparamagnetic iron oxide nanoparticles (SPIONs) have received increasing interest as contrast media in biomedical imaging and innovative therapeutic tools, in particular for loco-regional ablative treatments and drug delivery. The future of therapeutic applications would strongly benefit from improving the capability of the nanostructured constructs to reach the selected target, in particular beyond the intravascular space. Besides the decoration of SPIONs surface with ad hoc bioactive molecules, external magnetic fields are in principle able to remotely influence SPIONs' physiological biodistribution and concentrate them to a specific anatomical region or portion of a tissue. The reduction of SPIONs administered to the body and the need for defining the effective SPIONs local concentration suggest that PET/CT may be a method to quantitatively detect the nanoparticles accumulation in vivo at low concentration and assess their tridimensional distribution in response to an external magnetic field and in relation to the local anatomy highlighted by CT imaging. Here, we report on the possibility to assess the spatial distribution of magnetically-driven radiolabelled SPIONs in a peripheral tissue (mouse thigh) with microPET/CT imaging. To this aim we labelled SPIONs using
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
(Boc-aminooxy)acetic acid, ≥98.0% (T)