Merck
  • Prevalence of and Factors Associated With Multidrug Resistant Organism (MDRO) Colonization in 3 Nursing Homes.
Prevalence of and Factors Associated With Multidrug Resistant Organism (MDRO) Colonization in 3 Nursing Homes.
Infection control and hospital epidemiology (2016-09-28)
James A McKinnell, Loren G Miller, Raveena Singh, Ken Kleinman, Ellena M Peterson, Kaye D Evans, Tabitha D Dutciuc, Lauren Heim, Adrijana Gombosev, Marlene Estevez, Bryn Launer, Tom Tjoa, Steven Tam, Michael A Bolaris, Susan S Huang
RESUMO
Nursing home residents are at risk for acquiring and transmitting MDROs. A serial point-prevalence study of 605 residents in 3 facilities using random sampling found MDRO colonization in 45% of residents: methicillin-resistant Staphylococcus aureus (MRSA, 26%); extended-spectrum β-lactamase-producing Enterobacteriaceae (ESBL, 17%); vancomycin-resistant Enterococcus spp. (VRE, 16%); carbapenem-resistant Enterobacteriaceae (CRE, 1%). MDRO colonization was associated with history of MDRO, care needs, incontinence, and catheters. Infect Control Hosp Epidemiol 2016;1485-1488.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Cefpodoxime, VETRANAL®, analytical standard