Merck
eIF6 coordinates insulin sensitivity and lipid metabolism by coupling translation to transcription.
Nature communications (2015-09-19)
Daniela Brina, Annarita Miluzio, Sara Ricciardi, Kim Clarke, Peter K Davidsen, Gabriella Viero, Toma Tebaldi, Nina Offenhäuser, Jan Rozman, Birgit Rathkolb, Susanne Neschen, Martin Klingenspor, Eckhard Wolf, Valerie Gailus-Durner, Helmut Fuchs, Martin Hrabe de Angelis, Alessandro Quattrone, Francesco Falciani, Stefano Biffo
RESUMO
Insulin regulates glycaemia, lipogenesis and increases mRNA translation. Cells with reduced eukaryotic initiation factor 6 (eIF6) do not increase translation in response to insulin. The role of insulin-regulated translation is unknown. Here we show that reduction of insulin-regulated translation in mice heterozygous for eIF6 results in normal glycaemia, but less blood cholesterol and triglycerides. eIF6 controls fatty acid synthesis and glycolysis in a cell autonomous fashion. eIF6 acts by exerting translational control of adipogenic transcription factors like C/EBPβ, C/EBPδ and ATF4 that have G/C rich or uORF sequences in their 5' UTR. The outcome of the translational activation by eIF6 is a reshaping of gene expression with increased levels of lipogenic and glycolytic enzymes. Finally, eIF6 levels modulate histone acetylation and amounts of rate-limiting fatty acid synthase (Fasn) mRNA. Since obesity, type 2 diabetes, and cancer require a Fasn-driven lipogenic state, we propose that eIF6 could be a therapeutic target for these diseases.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Antiβ-actina monoclonal, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-HMG-CoA reductase Antibody, from rabbit, purified by affinity chromatography