Merck
  • Hepatocyte growth factor and c-Met in cervical intraepithelial neoplasia: overexpression of proteins associated with oncogenic human papillomavirus and human immunodeficiency virus.
Hepatocyte growth factor and c-Met in cervical intraepithelial neoplasia: overexpression of proteins associated with oncogenic human papillomavirus and human immunodeficiency virus.
Clinical cancer research : an official journal of the American Association for Cancer Research (2003-01-23)
Francine Walker, Stéphanie Kermorgant, Emile Daraï, Patrick Madelenat, Anne Claude Cremieux, Dominique Hénin, Thérèse Lehy
RESUMO
High prevalence of squamous cervical intraepithelial neoplasia (CIN) linked to oncogenic human papillomavirus (HPV) exits in HIV-infected women. Hepatocyte growth factor (HGF) and its receptor, c-Met, promote cell proliferation and are involved in tumor progression. Nothing is yet known about their expression in low- and high-grade CIN. Therefore, the expression, localization, and behavior of HGF and c-Met in normal and dysplastic cervical epithelium were investigated. We studied normal cervical mucosa from 10 healthy women, and low- and high-grade cervical lesions, uninfected (condyloma acuminata) or infected with oncogenic HPVs, from 40 HIV-negative and 48 HIV-positive women, using in situ molecular techniques, immunocytochemistry and morphoquantitative methods. In 154 oncogenic HPV-infected CIN encountered in biopsy samples, the total number of epithelial cell layers increased significantly during lesion progression. This number was significantly higher in HIV-positive than in HIV-negative women for CIN1 and CIN2 (P < 0.025 to P < 0.01). In HIV-negative women, the number and percentage of HGF and c-Met immunostained cell layers, and the intensity of immunostaining were enhanced in oncogenic HPV-infected lesions as compared with normal mucosa and condyloma acuminata. The latter parameters were significantly higher in tissues of HIV-positive women (oncogenic HPV-infected CIN1 and CIN2, normal-appearing mucosa contiguous to CIN, condyloma acuminata) than in the corresponding tissues of HIV-negative women (P < 0.025 to P < 0.0001). Overexpression of HGF/c-Met complex strongly correlates with oncogenic HPV and HIV infection. This overexpressed complex may stimulate cell proliferation in condyloma acuminata and participate in tumor progression in oncogenic HPV-infected lesions.