Merck
  • Bovine herpesvirus type-1 glycoprotein K (gK) interacts with UL20 and is required for infectious virus production.
Bovine herpesvirus type-1 glycoprotein K (gK) interacts with UL20 and is required for infectious virus production.
Virology (2016-09-24)
Muzammel Haque, Brent Stanfield, Konstantin G Kousoulas
RESUMO
We have previously shown that the HSV-1 gK and UL20 proteins interact and function in virion envelopment, membrane fusion, and neuronal entry. Alignment of the predicted secondary structures of gKs encoded by BoHV-1, HSV-1, HSV-2, EHV-1 and VZV indicated a high degree of domain conservation. Two BoHV-1 gK-null mutant viruses were created by either gK gene deletion or stop codon insertion. In addition, a V5 epitope-tag was inserted at the carboxyl terminus of gK gene to detect gK. The engineered gK-null mutant viruses failed to replicate and produce viral plaques. Co-immunoprecipitation of gK and UL20 expressed via different methods revealed that gK and UL20 physically interacted in the presence or absence of other viral proteins. Confocal microscopy showed that gK and UL20 colocalized in infected cells. These results indicate that BoHV-1 gK and UL20 may function in a similar manner to other alphaherpesvirus orthologues specified by HSV-1, PRV and EHV-1.
MATERIAIS
Número do produto
Marca
Descrição do produto
Nunc® Lab-Tek® II Chamber Slide system, wells 4, well area 1.7 cm2, sterile
Preço e disponibilidade não estão disponíveis no momento.