Merck
  • The use of valinomycin, nigericin and trichlorocarbanilide in control of the protonmotive force in Escherichia coli cells.
The use of valinomycin, nigericin and trichlorocarbanilide in control of the protonmotive force in Escherichia coli cells.
The Biochemical journal (1983-04-15)
S Ahmed, I R Booth
RESUMO
Valinomycin, nigericin and trichlorocarbanilide were assessed for their ability to control the protonmotive force in Escherichia coli cells. Valinomycin, at high K+ concentrations, was found to decrease the membrane potential delta phi and indirectly to decrease the pH gradient delta pH. Nigericin was found to have two modes of action. At low concentrations (0.05-2 microM) it carried out K+/H+ exchange and decreased delta pH. At higher concentrations (50 microM) it carried out a K+-dependent transfer of H+, decreasing both delta phi and delta pH. In EDTA-treated cells only the latter mode of action was evident, whereas in a mutant sensitive to deoxycholate both types of effect were observed. Trichlorocarbanilide is proposed as an alternative to nigericin for the specific control of delta pH, and it can be used in cells not treated with EDTA.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Nigericin sodium salt Ready Made Solution, 5 mg/mL (DMSO:ethanol 1:1)