Merck
  • Flt23k nanoparticles offer additive benefit in graft survival and anti-angiogenic effects when combined with triamcinolone.
Flt23k nanoparticles offer additive benefit in graft survival and anti-angiogenic effects when combined with triamcinolone.
Investigative ophthalmology & visual science (2012-03-20)
Yang Kyung Cho, Hironori Uehara, Jason R Young, Puneet Tyagi, Uday B Kompella, Xiaohui Zhang, Ling Luo, Nirbhai Singh, Bonnie Archer, Balamurali K Ambati
RESUMO
To determine if nanoparticles delivering plasmids expressing Flt23k (an anti-VEGF intraceptor) can enhance murine cornea transplant survival and whether their effect is synergistic with steroid therapy. Biodegradable PLGA Flt23k loaded or blank nanoparticles were prepared using the emulsion solvent evaporation Graft survival, corneal neovascularization, and corneal lymphangiogenesis were compared among the Flt23k nanoparticles, blank nanoparticles, triamcinolone acetonide, and PBS groups following subconjunctival injection in mice that underwent penetrating keratoplasty. Graft survival, corneal neovascularization, and corneal lymphangiogenesis in a group treated with both nanoparticles and steroid therapy were also analyzed. The Flt23k nanoparticle group showed less neovascularization, lymphangiogenesis, and graft failure compared with the PBS control group (P < 0.01). The 2-month graft survival rate was 20% in the Flt23k nanoparticle group with no grafts surviving in the PBS group. When the Flt23k nanoparticle was combined with steroid therapy, a significant increase in graft survival was seen compared with both steroid treatment alone (P < 0.05) and steroid combined with blank nanoparticle treatment (P < 0.05). The 2-month graft survival rate was 91.6% in the combination group compared with 47.6% in the triamcinolone-only group and 42.4% in the triamcinolone plus blank nanoparticle group. Anti-VEGF nanoparticles (Flt23k) have a significant effect on decreasing neovascularization and lymphangiogenesis, resulting in increased graft survival in penetrating keratoplasty. This beneficial effect is synergistically enhanced with steroid treatment.