Merck
  • A Single Dose-Escalation Study to Evaluate the Safety and Pharmacokinetics of Orally Administered Des-Aspartate Angiotensin I in Healthy Subjects.
A Single Dose-Escalation Study to Evaluate the Safety and Pharmacokinetics of Orally Administered Des-Aspartate Angiotensin I in Healthy Subjects.
Drugs in R&D (2016-09-30)
Ko-Onn Lee, Chin-Meng Khoo, Balram Chowbay, Yiong-Huak Chan, Meng-Kwoon Sim
RESUMO
Des-aspartate-angiotensin I (DAA-I) is an endogenous angiotensin peptide and a prototype angiotensin receptor agonist (ARA). It acts on the angiotensin AT