Merck
Convenient synthesis of palladium nanoparticles and catalysis of Hiyama coupling reaction in water.
Organic letters (2007-08-07)
Dipankar Srimani, Sudeshna Sawoo, Amitabha Sarkar
RESUMO
An efficient synthesis of Pd nanoparticles in water has been developed using a Fischer carbene complex of tungsten as the reductant and PEG as the capping agent. The colloidal palladium (1 mol %) efficiently catalyzes Hiyama cross-coupling reactions performed in air. Excellent yields of products were obtained with a wide range of substrates. Catalytic activity and stability of the nanoparticles were found to be inversely correlated.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Tungsten hexacarbonyl, 97%