Merck
  • Studies on anticoccidial agents. 13. Synthesis and anticoccidial activity of nitropyridine-2- and -3-sulfonamides and derivatives.
Studies on anticoccidial agents. 13. Synthesis and anticoccidial activity of nitropyridine-2- and -3-sulfonamides and derivatives.
Journal of medicinal chemistry (1980-12-01)
Y Morisawa, M Kataoka, H Nagahori, T Sakamoto, N Kitano, K Kusano, K Sato
RESUMO
Eight nitropyridinesulfonamides and pyridinesulfonamide N-oxides as their bioisosteres were prepared and evaluated for anticoccidial activity. Of these compounds, 2-, 4- and 5-nitropyridine-3-sulfonamides and pyridine-2- and -3-sulfonamide N-oxides were found to be active against Eimeria tenella. Thus, the relative positions, ortho or meta, of the substituents in nitropyridine-3-sulfonamides and pyridinesulfonamide N-oxides are important for anticoccidial activity. N-Substituted analogues of 5-nitropyridine-3-sulfonamide were also prepared and optimal anticoccidial activity was attained with the sulfonamide and its lower N-alkyl derivatives. The mode of action of 5-nitropyridine-3-sulfonamide was examined and found to be active in the sporozoite and the first schizogony stages.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
2-Chloro-4-nitropyridine, 97%