Merck
  • Bakuchiol sensitizes cancer cells to TRAIL through ROS- and JNK-mediated upregulation of death receptors and downregulation of survival proteins.
Bakuchiol sensitizes cancer cells to TRAIL through ROS- and JNK-mediated upregulation of death receptors and downregulation of survival proteins.
Biochemical and biophysical research communications (2016-04-02)
Mi Hee Park, Jong Han Kim, Young-Ho Chung, Seung Ho Lee
RESUMO
We investigated whether bakuchiol, an analog of resveratrol enhances the apoptosis ability of tumor necrosis factor (TNF)-related apoptosis-inducing ligand (TRAIL) in cancer cells. Bakuchiol enhanced expression of cell death receptor (DR) in TRAIL-sensitive and -resistant colon cancer cells in a dose-dependent manner. A combination of bakuchiol with TRAIL significantly inhibited cell growth of TRAIL sensitive HCT116 and TRAIL resistant HT-29 cells. The expression of TRAIL receptors; DR4 and DR5 was significantly increased by treatment of bakuchiol, however, the expression of survival proteins (e.g., cFLIP, survivin, XIAP and Bcl2) was suppressed. Moreover, the expression of apoptosis related proteins such as cleaved caspase-3, -8, -9 and PARP was increased by combination treatment of bakuchiol and TRAIL. Depletion of DR4 or DR5 by small interfering RNA significantly reversed the cell growth inhibitory effects of bakuchiol in HCT116 and HT-29 cells. Pretreatment with the c-Jun N-terminal kinase (JNK) inhibitor SP600125 and the reactive oxygen species (ROS) scavenger N-acetylcysteine reduced the bakuchiol induced cell growth inhibitory effects. The collective results suggest that bakuchiol facilitates TRAIL-induced apoptosis in colon cancer cells through up-regulation of the TRAIL receptors; DR4 and DR5 via ROS/JNK pathway signals.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
MISSION® esiRNA, targeting human TNFRSF10B