Merck
  • Judicious application of allyl protecting groups for the synthesis of 2-morpholin-4-yl-4-oxo-4H-chromen-8-yl triflate, a key precursor of DNA-dependent protein kinase inhibitors.
Judicious application of allyl protecting groups for the synthesis of 2-morpholin-4-yl-4-oxo-4H-chromen-8-yl triflate, a key precursor of DNA-dependent protein kinase inhibitors.
Organic letters (2006-12-15)
Sonsoles Rodriguez Aristegui, Marine Desage El-Murr, Bernard T Golding, Roger J Griffin, Ian R Hardcastle
RESUMO
[Structure: see text] 2-morpholin-4-yl-4-oxo-4H-chromen-8-yl 2,2,2-trifluoromethanesulfonate is a key intermediate for the synthesis of the DNA-dependent protein kinase (DNA-PK) inhibitor 8-dibenzothiophen-4-yl-2-morpholin-4-yl-chromen-4-one (NU7441). Two improved methods for the synthesis of this triflate have been developed: (A) in 35% overall yield, through modification of the published route, and (B) in 15% overall yield, by a new route employing a Baker-Venkataraman rearrangement to enable generation of the chromenone scaffold. Both syntheses depend on the judicious use of allyl protecting groups.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Tris(triphenylphosphine)rhodium(I) chloride, 99.9% trace metals basis