Merck
Single-step purification of full-length human androgen receptor.
Nuclear receptor signaling (2006-04-11)
Dalia Juzumiene, Ching-yi Chang, Daju Fan, Tanya Hartney, John D Norris, Donald P McDonnell
RESUMO
The full-length human androgen receptor with an N-terminal biotin acceptor peptide tag was overexpressed in Spodoptera frugiperda cells in the presence of 1 microM dihydrotestosterone. Site-specific biotinylation of BAP was achieved in vivo by co-expression of E. coli biotin holoenzyme synthetase. The androgen receptor was purified by single-step affinity chromatography using Streptavidin Mutein Matrix under native conditions. The resultant protein was active, stable, 95% homogeneous, and we obtained sufficient yield for use in functional and structural studies.
MATERIAIS
Número do produto
Marca
Descrição do produto
Roche
Streptavidin Mutein Matrix, suspension, pkg of 5 mL