Merck
  • Inhibition of the translocation and extracellular release of high-mobility group box 1 alleviates liver damage in fibrotic mice in response to D-galactosamine/lipopolysaccharide challenge.
Inhibition of the translocation and extracellular release of high-mobility group box 1 alleviates liver damage in fibrotic mice in response to D-galactosamine/lipopolysaccharide challenge.
Molecular medicine reports (2016-04-02)
Li Bai, Ming Kong, Qingfen Zheng, Xiaohui Zhang, Xin Liu, Kejia Zu, Yu Chen, Sujun Zheng, Junfeng Li, Feng Ren, Jinli Lou, Shuang Liu, Zhongping Duan
RESUMO
Acute liver injury in the setting of fibrosis is an area of interest in investigations, and remains to be fully elucidated. Previous studies have suggested the beneficial effects of liver fibrosis induced by thioacetamide and partial bile duct ligation against Fas‑mediated acute liver injury. The activation of AKT and extracellular signal-regulated kinase signaling is considered to be crucial in this hepatoprotection. To demonstrate the protection of CCl4‑induced liver fibrosis against lethal challenge, the present study compared the reactivity to lethal doses of D‑galactosamine (D-GalN)/lipopolysaccharide (LPS) between fibrotic mice and control mice groups. The extent of hepatic damage was assessed by survival rate and histopathological analysis. The molecular basis of the fibrosis‑based hepatoprotection was examined, with a particular focus on the translocation and release of high‑mobility group box (HMGB)1 and the inflammatory response triggered by HMGB1. Hepatoprotection induced by fibrosis was demonstrated by improved survival rates (100%, vs. 20%) and improved preservation of liver architecture in fibrotic mice subjected to D‑GalN/LPS, compared with control mice treated in the same way. D‑GalN/LPS evoked the translocation and release of HMGB1, detected by immunohistochemistry, in the control mice, which was significantly inhibited in the fibrotic mice. The gene expression levels of HMGB1‑associated proinflammatory cytokines, including interleukin (IL)‑1β, IL‑6, tumor necrosis factor‑α and IL‑12p40, were markedly inhibited in the fibrotic mice when exposed to D‑GalN/LPS. These findings confirmed that CCl4‑based fibrosis induced hepatoprotection, and provided evidence that fibrosis inhibited the translocation and release of HMGB1, and the proinflammatory response triggered by HMGB1. This alleviated liver damage following exposure to D‑GalN/LPS challenge.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Cy3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution