Merck
  • A robust analytical method for measurement of phytoestrogens and related metabolites in serum with liquid chromatography tandem mass spectrometry.
A robust analytical method for measurement of phytoestrogens and related metabolites in serum with liquid chromatography tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2016-01-28)
Hongmei Jiang, Xiangjun Liao, Carla M Wood, Chao-Wu Xiao, Yong-Lai Feng
RESUMO
A sensitive and robust method using high performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) was developed for quantitation of 13 phytoestrogens and related metabolites in rat serum samples. A new type of column, the Kinetex core-shell C18 column, was applied for rapid separation of the target analytes in 10min. Two enzymes, sulfatase H-1 and gulcuronidase H-5 from Helix pomatia were compared on the efficiency of releasing the conjugated forms of the target analytes to their free forms in serum samples. The method detection limit (MDL) defined as three times the signal to noise ratio in spiked serum matrix-based solutions was in the range of 0.1-3.5ng/mL. The linear dynamic calibration was in the broad range of 0.2-500ng/mL for all target compounds. Thirty-two rat serum samples from the rats that were fed with diets containing either casein or soy protein isolates with various amounts of isoflavones for 8 weeks were analyzed for the target analytes with the developed method. Nine target analytes were detected in the serum samples. Those detectable compounds are all the metabolites of the dietary isoflavones, suggesting that the diet isoflavones were mostly metabolized to their metabolites in rat.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Enterolactone, ~95% (HPLC)
Sigma-Aldrich
Enterodiol, ≥95.0% (HPLC)