Merck
  • Infection susceptibility and immune senescence with advancing age replicated in accelerated aging Lmna(Dhe) mice.
Infection susceptibility and immune senescence with advancing age replicated in accelerated aging Lmna(Dhe) mice.
Aging cell (2015-08-08)
Lijun Xin, Tony T Jiang, Jeremy M Kinder, James M Ertelt, Sing Sing Way
RESUMO
Aging confers increased susceptibility to common pathogens including influenza A virus. Despite shared vulnerability to infection with advancing age in humans and rodents, the relatively long time required for immune senescence to take hold practically restricts the use of naturally aged mice to investigate aging-induced immunological shifts. Here, we show accelerated aging Lmna(Dhe) mice with spontaneous mutation in the nuclear scaffolding protein, lamin A, replicate infection susceptibility, and substantial immune cell shifts that occur with advancing age. Naturally aged (≥ 20 month) and 2- to 3-month-old Lmna(Dhe) mice share near identically increased influenza A susceptibility compared with age-matched Lmna(WT) control mice. Increased mortality and higher viral burden after influenza infection in Lmna(Dhe) mice parallel reduced accumulation of lung alveolar macrophage cells, systemic expansion of immune suppressive Foxp3⁺ regulatory T cells, and skewed immune dominance among viral-specific CD8⁺T cells similar to the immunological phenotype of naturally aged mice. Thus, aging-induced infection susceptibility and immune senescence are replicated in accelerated aging Lmna(Dhe) mice.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anticorpo IgG de cabra anti-camundongo, F(a′b)2, conjugado com FITC, 1 mg/mL, Chemicon®