Merck
  • Reduced expression of ZDHHC2 is associated with lymph node metastasis and poor prognosis in gastric adenocarcinoma.
Reduced expression of ZDHHC2 is associated with lymph node metastasis and poor prognosis in gastric adenocarcinoma.
PloS one (2013-03-05)
Shu-Mei Yan, Jian-Jun Tang, Chun-Yu Huang, Shao-Yan Xi, Ma-Yan Huang, Jian-Zhong Liang, Yuan-Xue Jiang, Yu-Hong Li, Zhi-Wei Zhou, Ingemar Ernberg, Qiu-Liang Wu, Zi-Ming Du
RESUMO
Zinc finger, DHHC-type containing 2 (ZDHHC2), originally named as reduced expression associated with metastasis protein (REAM), has been proposed as a putative tumor/metastasis suppressor gene and is often aberrantly decreased in human cancers. However ZDHHC2 expression pattern and its clinical significance have not yet been investigated in gastric adenocarcinoma. Quantitative Real-Time PCR (qRT-PCR) and immunostaining were performed to detect ZDHHC2 expression in gastric adenocarcinoma, and then the correlation between ZDHHC2 expression and clinicpathologic parameters, and patient survival was analyzed. Compared to the adjacent normal tissues, ZDHHC2 expression was significantly reduced in gastric tumor tissues as shown by qRT-PCR and immunostaining. Low expression of ZDHHC2 was observed in 44.7% (211/472) of gastric adenocarcinoma patients, and was associated significantly with lymph node metastasis (p<0.001) and histological grade (p<0.001). Multivariate Cox regression analysis indicated that ZDHHC2 expression had a significant, independent predictive value for survival of gastric cancer patients (HR = 0.627, p = 0.001). Our data suggest that reduced ZDHHC2 expression is associated with lymph node metastasis and independently predicts an unfavorable prognosis in gastric adenocarcinoma patients.