Merck
  • Impaired expression of genes coding for reactive oxygen species scavenging enzymes in testes of Mtfr1/Chppr-deficient mice.
Impaired expression of genes coding for reactive oxygen species scavenging enzymes in testes of Mtfr1/Chppr-deficient mice.
Reproduction (Cambridge, England) (2007-08-22)
Massimiliano Monticone, Laura Tonachini, Sara Tavella, Paolo Degan, Roberta Biticchi, Fioretta Palombi, Rossella Puglisi, Carla Boitani, Ranieri Cancedda, Patrizio Castagnola
RESUMO
Mtfr1/Chppr is a nuclear gene coding for a mitochondrial protein capable of inducing fission of this organelle in a sequence-specific manner. Here we show that in mice, Mtfr1/Chppr is ubiquitously expressed and displays the highest level of expression in pubertal and adult testes and in particular in spermatids and Leydig cells. To investigate Mtfr1 function in vivo, we analyzed homozygous mice null for this gene obtained through a gene trap approach. We show that these mice fail to express Mtfr1 and that in their testes several genes coding for enzymes involved in the defense against oxidative stress are downregulated. Among these, we studied in particular glutathione peroxidase 3 and show its expression in selected testis cell types. Furthermore, we demonstrate oxidative DNA damage specifically in testes of Mtfr1-deficient mice likely resulting from a reduced antioxidant activity. As a whole, these data suggest that Mtfr1 protects the male gonads against oxidative stress.