Merck
  • Human phosphoribosylformylglycineamide amidotransferase (FGARAT): regional mapping, complete coding sequence, isolation of a functional genomic clone, and DNA sequence analysis.
Human phosphoribosylformylglycineamide amidotransferase (FGARAT): regional mapping, complete coding sequence, isolation of a functional genomic clone, and DNA sequence analysis.
Gene (1999-11-05)
D Patterson, J Bleskan, K Gardiner, J Bowersox
RESUMO
Purines play essential roles in many cellular functions, including DNA replication, transcription, intra- and extra-cellular signaling, energy metabolism, and as coenzymes for many biochemical reactions. The de-novo synthesis of purines requires 10 enzymatic steps for the production of inosine monophosphate (IMP). Defects in purine metabolism are associated with human diseases. Further, many anticancer agents function as inhibitors of the de-novo biosynthetic pathway. Genes or cDNAs for most of the enzymes comprising this pathway have been isolated from humans or other mammals. One notable exception is the phosphoribosylformylglycineamide amidotransferase (FGARAT) gene, which encodes the fourth step of this pathway. This gene has been cloned from numerous microorganisms and from Drosophila melanogaster and C. elegans. We report here the identification of a human cDNA containing the coding region of the FGARAT mRNA and the isolation of a P1 clone that contains an intact human FGARAT gene. The P1 clone corrects the purine auxotrophy and protein deficiency of Chinese hamster ovary (CHO) cell mutants (AdeB) deficient in both the activity and the protein for FGARAT. The P1 clone was used to regionally map the FGARAT gene to chromosome region 17p13, a location consistent with our prior assignment of this gene to chromosome 17. A comparison of the DNA sequence of the human FGARAT and FGARAT DNA sequence from 17 other organisms is reported. The isolation of this gene means that DNA clones for all the 10 steps of IMP synthesis have been isolated from humans or other mammals.