- Dehalogenation of Functionalized Alkyl Halides in Water at Room Temperature.
Dehalogenation of Functionalized Alkyl Halides in Water at Room Temperature.
Green chemistry : an international journal and green chemistry resource : GC (2014-12-23)
Nicholas A Isley, Matt S Hageman, Bruce H Lipshutz
PMID25530711
RESUMO
Alkyl bromides and chlorides can be reduced to the corresponding hydrocarbons utilizing zinc in the presence of an amine additive. The process takes place in water at ambient temperatures, enabled by a commercially available designer surfactant. The reaction medium can be readily recycled, and the amount of organic solvent invested for product isolation is minimal, leading to very low E Factors.