Merck
  • Multiple sclerosis autoantigen myelin basic protein escapes control by ubiquitination during proteasomal degradation.
Multiple sclerosis autoantigen myelin basic protein escapes control by ubiquitination during proteasomal degradation.
The Journal of biological chemistry (2014-04-18)
Alexey Belogurov, Anna Kudriaeva, Ekaterina Kuzina, Ivan Smirnov, Tatyana Bobik, Natalia Ponomarenko, Yelena Kravtsova-Ivantsiv, Aaron Ciechanover, Alexander Gabibov
RESUMO
The vast majority of cellular proteins are degraded by the 26S proteasome after their ubiquitination. Here, we report that the major component of the myelin multilayered membrane sheath, myelin basic protein (MBP), is hydrolyzed by the 26S proteasome in a ubiquitin-independent manner both in vitro and in mammalian cells. As a proteasomal substrate, MBP reveals a distinct and physiologically relevant concentration range for ubiquitin-independent proteolysis. Enzymatic deimination prevents hydrolysis of MBP by the proteasome, suggesting that an abnormally basic charge contributes to its susceptibility toward proteasome-mediated degradation. To our knowledge, our data reveal the first case of a pathophysiologically important autoantigen as a ubiquitin-independent substrate of the 26S proteasome.