Merck
  • Differential associations of circulating asymmetric dimethylarginine and cell adhesion molecules with metformin use in patients with type 2 diabetes mellitus and stable coronary artery disease.
Differential associations of circulating asymmetric dimethylarginine and cell adhesion molecules with metformin use in patients with type 2 diabetes mellitus and stable coronary artery disease.
Amino acids (2015-04-11)
Olga Kruszelnicka, Bernadeta Chyrchel, Alain Golay, Andrzej Surdacki
RESUMO
Metformin, the drug of first choice in type 2 diabetes mellitus (T2DM), reduces cardiovascular (CV) morbidity and mortality in part independently of improved glycemic control and changes in traditional risk factors. However, there are discordant reports on the effects of metformin on endothelial function in T2DM. Our aim was to compare biochemical endothelial markers in patients with stable coronary artery disease (CAD) and T2DM stratified by metformin use. We studied 70 patients (29 women, age 68 ± 9 years) with established T2DM referred for elective coronary angiography owing to stable angina who were receiving a standard CV medication and metformin or other oral antidiabetic drugs. Exclusion criteria included heart failure and other relevant coexistent disorders. Biochemical indices of endothelial dysfunction and activation at admission were compared according to metformin use for at least 1 year prior to index hospitalization. Clinical characteristics were similar in patients receiving metformin (n = 40) vs. those on other oral antidiabetic agents (n = 30). Plasma soluble vascular cell adhesion molecule-1 (sVCAM-1) was lower (553 ± 148 vs. 668 ± 170 µg/L, P = 0.004) and asymmetric dimethylarginine (ADMA) higher (0.53 ± 0.09 vs. 0.48 ± 0.08 µM, P = 0.01) in subjects on metformin, which was maintained in multivariate analysis. Symmetric dimethylarginine, intercellular adhesion molecule-1, monocyte chemotactic protein-1 and E-selectin did not differ across the groups. The results were substantially unchanged after exclusion of insulin users. Thus, metformin use appears differentially associated with sVCAM-1 and ADMA in patients with T2DM and stable CAD. Whether this observation may reflect different prognostic effects of these endothelial markers in diabetes remains to be studied.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
NG,NG′-Dimethyl-L-arginine di(p-hydroxyazobenzene-p′-sulfonate) salt, ≥99% (TLC)